Potassium phenoxymethyltrifluoroborate 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: Potassium trifluoro(phenoxymethyl)borate

H32030.03
H32030.03EA 206 EUR
H32030.03
Potassium phenoxymethyltrifluoroborate 95%
Potassium phenoxymethyltrifluoroborate

Purity: 95% CAS No.: 1027642-30-3 Molecular Formula: C7H7BF3KO Formula Weight: 214.04 MDL No.: MFCD10566516

Formel: C₇H₇BF₃KO
Molecular Weight: 214,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10566516
CAS-Nummer: 1027642-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR