2-Brom-3-chlor-5-(trifluormethyl)pyridin ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H32055.06
H32055.06EA
188
EUR
H32055.06
2-Brom-3-chlor-5-(trifluormethyl)pyridin ≥97%
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine
Purity: 97%
CAS No.: 75806-84-7
Molecular Formula: C6H2BrClF3N
Formula Weight: 260.44
Boiling Point: 80°/32mm
MDL No.: MFCD00153072
Formel:
C₆H₂BrClF₃N Siedepunkt: 80 °C (32 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153072 CAS-Nummer: 75806-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...