Ethyl-2-(trifluoromethyl)-4-methyl-5-pyrimidine carboxylate 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 4-methyl-2-(trifluoromethyl)pyrimidine-5-carboxylate

H32065.03
H32065.03EA 131 EUR
H32065.03
Ethyl-2-(trifluoromethyl)-4-methyl-5-pyrimidine carboxylate 97%
Ethyl-2-(trifluoromethyl)-4-methyl-5-pyrimidine carboxylate

Purity: 97% CAS No.: 306960-67-8 Molecular Formula: C9H9F3N2O2 Formula Weight: 234.18 Boiling Point: 54°/0.5mm MDL No.: MFCD07367386

Formel: C₉H₉F₃N₂O₂
Molecular Weight: 234,18 g/mol
Siedepunkt: 54 °C (0,5 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD07367386
CAS-Nummer: 306960-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR