N-Boc-4-carboxymethoxypiperidine 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
N-Boc-4-Carboxymethoxypiperidine, (1-Boc-4-piperidinyloxy)acetic acid
H32102.03
H32102.03EA
124
EUR
H32102.03
N-Boc-4-carboxymethoxypiperidine 95%
N-Boc-4-carboxymethoxypiperidine
Purity: 95%
CAS No.: 161948-70-5
Molecular Formula: C12H21NO6
Formula Weight: 259.30
Melting Point: 105-107°
MDL No.: MFCD02178987
Formel:
C₁₂H₂₁NO₅ Schmelzpunkt: 105…107 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02178987 CAS-Nummer: 161948-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...