5-Methyl-3-phenylisoxazole-4-carboxaldehyde 97%
Lieferant: Thermo Scientific
|
Achtung
|
H32129.03
H32129.03EA
77.1
EUR
H32129.03
5-Methyl-3-phenylisoxazole-4-carboxaldehyde 97%
5-Methyl-3-phenylisoxazole-4-carboxaldehyde
Purity: 97%
CAS No.: 87967-95-1
Molecular Formula: C11H9NO2
Formula Weight: 187.20
Storage Temperature: 2°C to 8°C
Melting Point: 51-53°
MDL No.: MFCD02677714
Formel:
C₁₁H₉NO₂ Molecular Weight: 187,20 g/mol Schmelzpunkt: 51…53 °C |
MDL:
MFCD02677714 CAS-Nummer: 87967-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...