3-Methylpyridine-4-boronic acid pinacol ester 95%

Lieferant: Thermo Fisher Scientific
Achtung

H32135.MD
H32135.MDEA 397 EUR
H32135.MD
3-Methylpyridine-4-boronic acid pinacol ester 95%
3-Methylpyridine-4-boronic acid pinacol ester

Purity: 95% CAS No.: 1032358-00-1 Molecular Formula: C12H18BNO2 Formula Weight: 219.09 Storage Temperature: 2°C to 8°C MDL No.: MFCD07374972

Formel: C₁₂H₁₈BNO₂
MDL: MFCD07374972
CAS-Nummer: 1032358-00-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR