3-Chlor-5-fluorphenylboronsäure 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Chloro-5-fluorobenzeneboronic acid, 3-Chloro-5-fluorophenlyboronic acid

H32137.03 H32137.06
H32137.03EA 109 EUR
H32137.03 H32137.06
3-Chlor-5-fluorphenylboronsäure 95%
3-Chlor-5-fluorphenylboronsäure

Purity: 95% CAS No.: 328956-61-2 Molecular Formula: C6H5BClFO2 Formula Weight: 174.37 MDL No.: MFCD06801741

Formel: C₆H₅BClFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06801741
CAS-Nummer: 328956-61-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR