3,5-Dimethylpyridine-4-boronic acid pinacol ester 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

H32159.ME
H32159.MEEA 324 EUR
H32159.ME
3,5-Dimethylpyridine-4-boronic acid pinacol ester 95%
3,5-Dimethylpyridine-4-boronic acid pinacol ester

Purity: 95% CAS No.: 1032358-02-3 Molecular Formula: C13H20BNO2 Formula Weight: 233.12 Storage Temperature: 2°C to 8°C MDL No.: MFCD10566506

Formel: C₁₃H₂₀BNO₂
Molecular Weight: 233,12 g/mol
MDL: MFCD10566506
CAS-Nummer: 1032358-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR