Kalium(brommethyl)trifluoroborat 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(Brommethyl)trifluorborsäure Kaliumsalz
H32168.06
H32168.06EA
198
EUR
H32168.06
Kalium(brommethyl)trifluoroborat 95%
Kalium(brommethyl)trifluoroborat
Purity: 95%
CAS No.: 888711-44-2
Molecular Formula: CH2BBrF3K
Formula Weight: 200.84
MDL No.: MFCD09265154
Formel:
CH₂BBrF₃K Molecular Weight: 200,83 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09265154 CAS-Nummer: 888711-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...