3-Chloro-4-(pyridin-3-yl)-1,2,5-thiadiazole 95%

Lieferant: Thermo Scientific
Achtung

H32169.03
H32169.03EA 197 EUR
H32169.03
3-Chloro-4-(pyridin-3-yl)-1,2,5-thiadiazole 95%
3-Chloro-4-(pyridin-3-yl)-1,2,5-thiadiazole

Purity: 95% CAS No.: 131986-28-2 Molecular Formula: C7H4ClN3S Formula Weight: 197.65 Melting Point: 56° MDL No.: MFCD03066187

Formel: C₇H₄ClN₃S
Schmelzpunkt: 56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03066187
CAS-Nummer: 131986-28-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR