5-Fluoroxindol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Fluoroindolin-2-one, 5-Fluoro-2-oxoindoline, 5-Fluoro-2-oxindole
H32176.03
H32176.03EA
103
EUR
H32176.03
5-Fluoroxindol ≥97%
5-Fluoroxindol
Purity: 97%
CAS No.: 56341-41-4
Molecular Formula: C8H6FNO
Formula Weight: 151.14
Melting Point: 142-146°
MDL No.: MFCD02179598
Formel:
C₈H₆FNO Molecular Weight: 151,14 g/mol Schmelzpunkt: 142…146 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179598 CAS-Nummer: 56341-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...