α,3-Dibrom-2-fluortoluol ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-Bromo-3-(bromomethyl)-2-fluorobenzene, 3-Bromo-2-fluorobenzyl bromide
H32228.06
H32228.06EA
236
EUR
H32228.06
α,3-Dibrom-2-fluortoluol ≥97%
α,3-Dibrom-2-fluortoluol
Purity: 97%
CAS No.: 149947-16-0
Molecular Formula: C7H5Br2F
Formula Weight: 267.92
Boiling Point: 60°/0.1mm
UN No.: 3265
MDL No.: MFCD09842442
Formel:
C₇H₅Br₂F Molecular Weight: 267,92 g/mol Siedepunkt: 60 °C (0,1 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD09842442 CAS-Nummer: 149947-16-0 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...