2,3-Difluor-4-hydroxybenzonitril 95%

Lieferant: Thermo Scientific
Achtung

H32231.03
H32231.03EA 69 EUR
H32231.03
2,3-Difluor-4-hydroxybenzonitril 95%
2,3-Difluor-4-hydroxybenzonitril

Purity: 95% CAS No.: 126162-38-7 Molecular Formula: C7H3F2NO Formula Weight: 155.10 Melting Point: 145-149° MDL No.: MFCD00269596

Formel: C₇H₃F₂NO
Molecular Weight: 155,1 g/mol
Schmelzpunkt: 145…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00269596
CAS-Nummer: 126162-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR