3-Amino-1-(4-chlorophenyl)-1H-pyrazole 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(4-Chloro-phenyl)-1H-pyrazol-3-ylamine
H32252.MD
H32252.MDEA
188
EUR
H32252.MD
3-Amino-1-(4-chlorophenyl)-1H-pyrazole 95%
3-Amino-1-(4-chlorophenyl)-1H-pyrazole
Purity: 95%
CAS No.: 66000-39-3
Molecular Formula: C9H8ClN3
Formula Weight: 193.64
Melting Point: 107-108°
MDL No.: MFCD09027932
Formel:
C₉H₈ClN₃ Molecular Weight: 193,64 g/mol Schmelzpunkt: 107…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027932 CAS-Nummer: 66000-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...