1,1'-Diethyl-2,2'-carbocyanine bromide 95%
Lieferant: Thermo Scientific
|
Achtung
|
H32253.03
H32253.03EA
51.6
EUR
H32253.03
1,1'-Diethyl-2,2'-carbocyanine bromide 95%
1,1'-Diethyl-2,2'-carbocyanine bromide
Purity: 95%
CAS No.: 2670-67-9
Molecular Formula: C25H25BrN2
Formula Weight: 433.39
MDL No.: MFCD00011973
Formel:
C₂₅H₂₅BrN₂ Molecular Weight: 433,39 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011973 CAS-Nummer: 2670-67-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...