Ethylamin 2.0 M in THF
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H32306.AE
H32306.AP
H32306.AEEA
64.5
EUR
H32306.AE
H32306.AP
Ethylamin 2.0 M in THF
Ethylamin
Formel:
C₂H₇N Dichte: 0,856 Flammpunkt: –34 °C (–29 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008160 CAS-Nummer: 75-04-7 EINECS: 200-834-7 UN: 2056 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...