2-Ethyl-5,5-dimethyl-1,3-dioxane 1000 mg/l in Acetonitril

Lieferant: Thermo Scientific
Gefahr

H32308.AE
H32308.AEEA 230 EUR
H32308.AE
2-Ethyl-5,5-dimethyl-1,3-dioxane 1000 mg/l in Acetonitril
2-Ethyl-5,5-dimethyl-1,3-dioxane

UN No.: UN1648 MDL No.: MFCD16622268

Formel: C₈H₁₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16622268
CAS-Nummer: 768-58-1
UN: 1648
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR