5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Bromo-2-(trifluoromethyl)isonicotinic acid

H32316.03
H32316.03EA 168 EUR
H32316.03
5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid 95%
5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid

Purity: 95% CAS No.: 749875-16-9 Molecular Formula: C7H3BrF3NO2 Formula Weight: 270.00 Melting Point: 235-238° MDL No.: MFCD10687236

Formel: C₇H₃BrF₃NO₂
Molecular Weight: 270,01 g/mol
Schmelzpunkt: 235…238 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD10687236
CAS-Nummer: 749875-16-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR