2-tert-Butyl-4-methylthiazole-5-methanol 97%
Lieferant: Thermo Scientific
H32329.06
H32329.06EA
220
EUR
H32329.06
2-tert-Butyl-4-methylthiazole-5-methanol 97%
2-tert-Butyl-4-methylthiazole-5-methanol
Purity: 97%
Molecular Formula: C9H15NOS
Formula Weight: 185.29
Flash Point:
Learn more