2-tert-Butyl-4-methylthiazole-5-methanol 97%
Lieferant: Thermo Scientific
H32329.06
H32329.06EA 220 EUR
H32329.06
2-tert-Butyl-4-methylthiazole-5-methanol 97%
2-tert-Butyl-4-methylthiazole-5-methanol
Purity: 97% Molecular Formula: C9H15NOS Formula Weight: 185.29 Flash Point: 100°(212°F) Refractive Index: 1.5250 Boiling Point: 110°/0.75mm MDL No.: MFCD16660039