4-(Trifluormethyl)phenylhydraziniumchlorid 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-(4-(Trifluormethyl)phenyl)hydrazin Hydrochlorid, 4-(Trifluormethyl)phenylhydrazin Hydrochlorid, 4-Hydrazinobenzotrifluorid Hydrochlorid, 4-(Trifluormethyl)phenylhydrazinhydrochlorid
H32340.03
H32340.09
H32340.03EA
32.5
EUR
H32340.03
H32340.09
4-(Trifluormethyl)phenylhydraziniumchlorid 96%
4-(Trifluormethyl)phenylhydraziniumchlorid
Purity: 96%
CAS No.: 2923-56-0
Molecular Formula: C7H7F3N2.HCl
Formula Weight: 212.60
Melting Point: ca 210° dec.
MDL No.: MFCD00204233
Formel:
C₇H₈ClF₃N₂ Schmelzpunkt: ∼ 210 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00204233 CAS-Nummer: 2923-56-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...