2-Nitro-4-pyridinecarboxylic acid 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Nitroisonicotinic acid
H32361.03
H32361.03EA
148
EUR
H32361.03
2-Nitro-4-pyridinecarboxylic acid 97%
2-Nitro-4-pyridinecarboxylic acid
Purity: 97%
CAS No.: 33225-74-0
Molecular Formula: C6H4N2O4
Formula Weight: 168.11
Melting Point: ca 170° dec.
MDL No.: MFCD04114160
Formel:
C₆H₄N₂O₄ Molecular Weight: 168,11 g/mol Schmelzpunkt: ∼ 170 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114160 CAS-Nummer: 33225-74-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...