5-Brom-6-chlornicotinsäure 97%

Lieferant: Thermo Scientific
Achtung

H32384.06
H32384.06EA 161 EUR
H32384.06
5-Brom-6-chlornicotinsäure 97%
5-Bromo-6-chloronicotinic acid

Purity: 97% CAS No.: 29241-62-1 Molecular Formula: C6H3BrClNO2 Formula Weight: 236.45 Melting Point: 170-173° MDL No.: MFCD01927098

Formel: C₆H₃BrClNO₂
Molecular Weight: 236,45 g/mol
Schmelzpunkt: 170…173 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01927098
CAS-Nummer: 29241-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR