Tetrafluorpyrimidin 95%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2,4,5,6-Tetrafluoropyrimidine

H32453.MD
H32453.MDEA 169 EUR
H32453.MD
Tetrafluorpyrimidin 95%
Tetrafluorpyrimidin

Purity: 95% CAS No.: 767-79-3 Molecular Formula: C4F4N2 Formula Weight: 152.05 Boiling Point: 83° UN No.: 3265 MDL No.: MFCD10687402

Formel: C₄F₄N₂
Siedepunkt: 83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD10687402
CAS-Nummer: 767-79-3
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR