Amberlite® IRA-96 Ionenaustauscherharz
Lieferant: Thermo Fisher Scientific
H32475.0B
H32475.30
H32475.0BEA
76.6
EUR
H32475.0B
H32475.30
Amberlite® IRA-96 Ionenaustauscherharz
Amberlite® IRA-96
CAS No.: 39409-19-3
Physical Form: Beads
Appearance: Pale brown
MDL No.: MFCD00166430
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD00166430 CAS-Nummer: 39409-19-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...