4-(Aminomethyl)pyrimidine hydrochloride 97%

Lieferant: Thermo Scientific
Achtung

H32582.ME
H32582.MEEA 196 EUR
H32582.ME
4-(Aminomethyl)pyrimidine hydrochloride 97%
4-(Aminomethyl)pyrimidine hydrochloride

Purity: 97% CAS No.: 45588-79-2 Molecular Formula: C5H7N3.HCl Formula Weight: 145.59 Melting Point: 176-178° MDL No.: MFCD07375466

Formel: C₅H₇N₃·HCl
Molecular Weight: 145,59 g/mol
Schmelzpunkt: 176…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07375466
CAS-Nummer: 1138011-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR