Ammonium O,O'-dimethyldithiophosphate 95%

Lieferant: Thermo Fisher Scientific
Achtung

H32592.03 H32592.09
H32592.03EA 38.1 EUR
H32592.03 H32592.09
Ammonium O,O'-dimethyldithiophosphate 95%
Ammonium O,O'-dimethyldithiophosphate

Purity: 95% CAS No.: 1066-97-3 Molecular Formula: C2H10NO2PS2 Formula Weight: 175.21 Odour: Weak Melting Point: 146-150° MDL No.: MFCD09753116

Formel: C₂H₁₀NO₂PS₂
Schmelzpunkt: 146…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09753116
CAS-Nummer: 1066-97-3
EINECS: 213-922-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR