Ammonium O,O'-dimethyldithiophosphate 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H32592.03
H32592.09
H32592.03EA
38.1
EUR
H32592.03
H32592.09
Ammonium O,O'-dimethyldithiophosphate 95%
Ammonium O,O'-dimethyldithiophosphate
Purity: 95%
CAS No.: 1066-97-3
Molecular Formula: C2H10NO2PS2
Formula Weight: 175.21
Odour: Weak
Melting Point: 146-150°
MDL No.: MFCD09753116
Formel:
C₂H₁₀NO₂PS₂ Schmelzpunkt: 146…150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09753116 CAS-Nummer: 1066-97-3 EINECS: 213-922-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...