5-Chlor-2-pyrazincarbonsäure 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H32596.03
H32596.03EA
91.4
EUR
H32596.03
5-Chlor-2-pyrazincarbonsäure 95%
5-Chloropyrazine-2-carboxylic acid
Purity: 95%
CAS No.: 36070-80-1
Molecular Formula: C5H3ClN2O2
Formula Weight: 158.54
Physical Form: Solid
Melting Point: 150-160°
MDL No.: MFCD09033269
Formel:
C5H3ClN2O2 Schmelzpunkt: 150…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09033269 CAS-Nummer: 36070-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...