4-Hydroxy-3-(trifluoromethoxy)benzaldehyde ≥98%

Lieferant: Thermo Scientific
Achtung

H32602.03
H32602.03EA 87.5 EUR
H32602.03
4-Hydroxy-3-(trifluoromethoxy)benzaldehyde ≥98%
4-Hydroxy-3-(trifluoromethoxy)benzaldehyde

Purity: 98+% CAS No.: 53104-95-3 Molecular Formula: C8H5F3O3 Formula Weight: 206.12 Melting Point: 76-80° MDL No.: MFCD04973780

Formel: C₈H₅F₃O₃
Molecular Weight: 206,12 g/mol
Schmelzpunkt: 76…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04973780
CAS-Nummer: 53104-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR