5-Amino-2,4-dichlorpyrimidin 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,4-Dichloropyrimidin-5-amine
H32606.06
H32606.06EA
279
EUR
H32606.06
5-Amino-2,4-dichlorpyrimidin 97%
5-Amino-2,4-dichloropyrimidine
Purity: 97%
CAS No.: 5177-27-5
Molecular Formula: C4H3Cl2N3
Formula Weight: 164.00
MDL No.: MFCD05662684
Formel:
C₄H₃Cl₂N₃ Molecular Weight: 164,00 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05662684 CAS-Nummer: 5177-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...