4-Iod-1-phenyl-1H-pyrazol 95%

Lieferant: Thermo Fisher Scientific
Achtung

H32612.MD H32612.03
H32612.MDEA 114 EUR
H32612.MD H32612.03
4-Iod-1-phenyl-1H-pyrazol 95%
4-Iodo-1-phenyl-1H-pyrazole

Purity: 95% CAS No.: 23889-85-2 Molecular Formula: C9H7IN2 Formula Weight: 270.07 MDL No.: MFCD11707168

Formel: C₉H₇IN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11707168
CAS-Nummer: 23889-85-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR