4-Brom-2-(trifluormethoxy)anisol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H32645.03 H32645.06
H32645.03EA 38.3 EUR
H32645.03 H32645.06
4-Brom-2-(trifluormethoxy)anisol ≥98%
4-Brom-2-(trifluormethoxy)anisol

Purity: 98% CAS No.: 853771-88-7 Molecular Formula: C8H6BrF3O2 Formula Weight: 271.04 MDL No.: MFCD09907950

Formel: C₈H₆BrF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09907950
CAS-Nummer: 853771-88-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR