2-Chlor-7-methoxy-4-methylchinolin ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 7-Methoxy-2-chloro-4-methylquinoline

H32685.03
H32685.03EA 71.9 EUR
H32685.03
2-Chlor-7-methoxy-4-methylchinolin ≥97%
2-Chloro-7-methoxy-4-methylquinoline

Purity: 97% CAS No.: 97892-67-6 Molecular Formula: C11H10ClNO Formula Weight: 207.66 Physical Form: Crystalline solid Appearance: White to pale brown Melting Point: 111-113° MDL No.: MFCD00711757

Formel: C₁₁H₁₀ClNO
Molecular Weight: 207,66 g/mol
Schmelzpunkt: 111…113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00711757
CAS-Nummer: 97892-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR