3-Brom-5-fluorbenzoesäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
H32692.06
H32692.06EA
296
EUR
H32692.06
3-Brom-5-fluorbenzoesäure ≥97%
3-Bromo-5-fluorobenzoic acid
Purity: 97%
CAS No.: 176548-70-2
Molecular Formula: C7H4BrFO2
Formula Weight: 219.01
Melting Point: 141-145°
MDL No.: MFCD01569540
Formel:
C₇H₄BrFO₂ Molecular Weight: 219,01 g/mol Schmelzpunkt: 141…145 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01569540 CAS-Nummer: 176548-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...