1-Boc-DL-nipecotinsäureethylester ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Ethyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate, Ethyl 1-Boc-DL-nipecotate, Ethyl 1-Boc-3-piperidinecarboxylate

H32704.06
H32704.06EA 116 EUR
H32704.06
1-Boc-DL-nipecotinsäureethylester ≥97%
1-Boc-DL-nipecotinsäureethylester

Purity: 97% CAS No.: 130250-54-3 Molecular Formula: C13H23NO4 Formula Weight: 257.33 Melting Point: 31-35° Boiling Point: 95°/0.5mm MDL No.: MFCD04116274

Formel: C₁₃H₂₃NO₄
Siedepunkt: 95 °C (0,5 mmHg)
Schmelzpunkt: 31…35 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04116274
CAS-Nummer: 130250-54-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR