2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,2,2-Trifluoro-1-(pyridin-3-yl)ethanamine hydrochloride
H32717.03
H32717.03EA
161
EUR
H32717.03
2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride 95%
2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride
Purity: 95%
Molecular Formula: C7H7F3N2.HCl
Formula Weight: 212.60
MDL No.: MFCD06739079
Formel:
C₇H₇F₃N₂·HCl Molecular Weight: 212,60 g/mol Schmelzpunkt: 97…102 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06739079 CAS-Nummer: 1138011-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...