2-Fluor-5-methoxybenzonitril ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H32724.06
H32724.06EA
69.5
EUR
H32724.06
2-Fluor-5-methoxybenzonitril ≥98%
2-Fluor-5-methoxybenzonitril
Purity: 98%
CAS No.: 127667-01-0
Molecular Formula: C8H6FNO
Formula Weight: 151.14
Melting Point: 81-85°
UN No.: 3439
MDL No.: MFCD00142921
Formel:
C₈H₆FNO Schmelzpunkt: 81…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00142921 CAS-Nummer: 127667-01-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...