3-(4-Biphenyl)propionic acid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-([1,1'-Biphenyl]-4-yl)propanoic acid

H32729.06
H32729.06EA 160 EUR
H32729.06
3-(4-Biphenyl)propionic acid 98%
3-(4-Biphenyl)propionic acid

Purity: 98% CAS No.: 35888-99-4 Molecular Formula: C15H14O2 Formula Weight: 226.28 Melting Point: 149-152° MDL No.: MFCD06823979

Formel: C₁₅H₁₄O₂
Molecular Weight: 226,275 g/mol
Schmelzpunkt: 149…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06823979
CAS-Nummer: 35888-99-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR