3-Methylbenzoesäureanhydrid ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H32739.03 H32739.06
H32739.03EA 31.9 EUR
H32739.03 H32739.06
3-Methylbenzoesäureanhydrid ≥97%
3-Methylbenzoic anhydride

Purity: 97% CAS No.: 21436-44-2 Molecular Formula: C16H14O3 Formula Weight: 254.29 Melting Point: 66-70° MDL No.: MFCD00048083

Formel: C₁₆H₁₄O₃
Schmelzpunkt: 66…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00048083
CAS-Nummer: 21436-44-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR