Methyl-2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate

H32755.06 H32755.14
H32755.06EA 68 EUR
H32755.06 H32755.14
Methyl-2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate 98%
Methyl-2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Purity: 98% CAS No.: 108354-78-5 Molecular Formula: C10H13NO2S Formula Weight: 211.28 Melting Point: 126-129° MDL No.: MFCD00130099

Formel: C₁₀H₁₃NO₂S
Molecular Weight: 211.28 g/mol
Schmelzpunkt: 126…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00130099
CAS-Nummer: 108354-78-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR