3,5-Di-tert-butylanilin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H32768.03
H32768.03EA 125 EUR
H32768.03
3,5-Di-tert-butylanilin ≥97%
3,5-Di-tert-butylanilin

Purity: 97% CAS No.: 2380-36-1 Molecular Formula: C14H23N Formula Weight: 205.35 Melting Point: 54-57° MDL No.: MFCD00134096

Formel: C₁₄H₂₃N
Schmelzpunkt: 54…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00134096
CAS-Nummer: 2380-36-1
EINECS: 219-173-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR