2-Nitro-4-(trifluormethyl)benzylbromid ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H32802.03
H32802.06
H32802.03EA
66.1
EUR
H32802.03
H32802.06
2-Nitro-4-(trifluormethyl)benzylbromid ≥97%
2-Nitro-4-(trifluormethyl)benzylbromid
Purity: 97%
CAS No.: 162333-02-0
Molecular Formula: C8H5BrF3NO3
Formula Weight: 284.03
Physical Form: Solid
Appearance: White to brown
Odour: Pungent
Melting Point: 36-39°
UN No.: 3261
MDL No.: MFCD03095413
Formel:
C₈H₅BrF₃NO₂ Schmelzpunkt: 36…39 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095413 CAS-Nummer: 162333-02-0 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...