2-Amino-5-phenyl-1,3,4-thiadiazole 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Amino-5-phenyl-1,3,4-thiadiazol, 5-Phenyl-1,3,4-thiadiazol-2-amine
H32826.03
H32826.03EA
33.4
EUR
H32826.03
2-Amino-5-phenyl-1,3,4-thiadiazole 97%
2-Amino-5-phenyl-1,3,4-thiadiazole
Purity: 97%
CAS No.: 2002-03-1
Molecular Formula: C8H7N3S
Formula Weight: 177.23
Melting Point: 227-230°
MDL No.: MFCD00205278
Formel:
C₈H₇N₃S Molecular Weight: 177,23 g/mol Schmelzpunkt: 227…230 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00205278 CAS-Nummer: 2002-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...