6-Hydroxy-1-tetralon ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H32840.06
H32840.06EA
69.6
EUR
H32840.06
6-Hydroxy-1-tetralon ≥97%
6-Hydroxy-1-tetralon
Purity: 97%
CAS No.: 3470-50-6
Molecular Formula: C10H10O2
Formula Weight: 162.19
Melting Point: 154-157°
MDL No.: MFCD00156670
Formel:
C₁₀H₁₀O₂ Molecular Weight: 162.19 g/mol Schmelzpunkt: 154…157 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00156670 CAS-Nummer: 3470-50-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...