Ethyl-1-(trifluoromethyl)cyclobutanecarboxylate 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(Trifluoromethyl)cyclobutanecarboxylic acid ethyl ester, Ethyl 1-(trifluoromethyl)cyclobutanecarboxylate
H32847.03
H32847.03EA
125
EUR
H32847.03
Ethyl-1-(trifluoromethyl)cyclobutanecarboxylate 97%
Ethyl-1-(trifluoromethyl)cyclobutanecarboxylate
Purity: 97%
Molecular Formula: C8H11F3O2
Formula Weight: 196.17
Boiling Point: 61-63°/20mm
MDL No.: MFCD10699116
Formel:
C₈H₁₁F₃O₂ Molecular Weight: 196,17 g/mol Siedepunkt: 61…63 °C (20 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD10699116 CAS-Nummer: 1040683-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...