6-Fluor-3-formylchromon ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Fluorochromone-3-carboxaldehyde, 6-Fluoro-4-oxo-4H-chromene-3-carbaldehyde

H32857.03
H32857.03EA 26.7 EUR
H32857.03
6-Fluor-3-formylchromon ≥97%
6-Fluor-3-formylchromon

Purity: 97% CAS No.: 69155-76-6 Molecular Formula: C10H5FO3 Formula Weight: 192.15 Melting Point: 155-160° MDL No.: MFCD00139060

Formel: C₁₀H₅FO₃
Molecular Weight: 192,15 g/mol
Schmelzpunkt: 155…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00139060
CAS-Nummer: 69155-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR