6-Brom-m-toluylsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H32927.03 H32927.06
H32927.03EA 21.2 EUR
H32927.03 H32927.06
6-Brom-m-toluylsäure ≥98%
6-Brom-m-toluylsäure

Purity: 98% CAS No.: 6967-82-4 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 MDL No.: MFCD00079722

Formel: C₈H₇BrO₂
Molecular Weight: 215.05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00079722
CAS-Nummer: 6967-82-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR