Ethyl-2-fluoroisobutyrate 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Ethyl 2-fluoro-2-methylpropanoate

H32931.06
H32931.06EA 121 EUR
H32931.06
Ethyl-2-fluoroisobutyrate 97%
Ethyl-2-fluoroisobutyrate

Purity: 97% CAS No.: 55816-69-8 Molecular Formula: C6H11FO2 Formula Weight: 134.15 Boiling Point: 122-126° UN No.: UN1992 MDL No.: MFCD08062432

Formel: C₆H₁₁FO₂
Molecular Weight: 134,15 g/mol
Siedepunkt: 122…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08062432
CAS-Nummer: 55816-69-8
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR