Methyl-1H-indazol-6-carboxylat 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Methyl 1H-indazole-6-carboxylate, Indazole-6-carboxylic acid methyll ester, Methyl Indazole-6-carboxylate
H32933.03
H32933.03EA
141
EUR
H32933.03
Methyl-1H-indazol-6-carboxylat 95%
Methyl-1H-indazol-6-carboxylat
Purity: 95%
CAS No.: 170487-40-8
Molecular Formula: C9H8N2O2
Formula Weight: 176.17
Melting Point: 142-144°
MDL No.: MFCD07371612
Formel:
C₉H₈N₂O₂ Molecular Weight: 176,17 g/mol Schmelzpunkt: 142…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07371612 CAS-Nummer: 170487-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...