2-Methyl-1,3-benzothiazol-5-ol 97%

Lieferant: Thermo Fisher Scientific
Achtung

H32937.03 H32937.09
H32937.03EA 36.1 EUR
H32937.03 H32937.09
2-Methyl-1,3-benzothiazol-5-ol 97%
2-Methyl-1,3-benzothiazol-5-ol

Purity: 97% CAS No.: 68867-14-1 Molecular Formula: C8H7NOS Formula Weight: 165.21 Melting Point: 185-187° MDL No.: MFCD00192276

Formel: C₈H₇NOS
Schmelzpunkt: 185…187 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00192276
CAS-Nummer: 68867-14-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR